Description
Capture and isolate contaminants to ensure the delivery of unadulterated cleaning and sanitizing agents. No more re-applying dirty water and weakened solutions. TruCLEAN Pro systems efficiently clean and disinfect floors, walls, and ceilings in large or small area cleanrooms of any grade.
The adaptable TruCLEAN Pro system allows triple (15 L) or double (22 L) bucket configuration in a heavy-duty stainless-steel cart designed for strength, durability, and hygiene. Our Pro series offers the most versatile mopping equipment with multiple configurations, colorways, and accessories to customize for your specific environment.
Each mopping system component plays an important role in delivering the highest level of cleanliness. For example, our innovative swivel mop frame is accurately balanced to maintain total surface contact at any angle. The unique wringing mechanism exerts force evenly across the entire mop surface, extracting the maximum amount of soiled contaminants.
Specifications
Specifications
| Color | Green |
| Dimensions (L x W x H) | 27 x 19 x 41 in. |