Description
Capture and isolate contaminants to ensure the delivery of unadulterated cleaning and sanitizing agents. No more re-applying dirty water and weakened solutions. The Pro XL system is designed to tackle very large area cleaning of floors, walls, and ceilings.
Equipped with large 22 Liter buckets and two 6 Liter pails in a handle-mounted carrying basket, the XL system easily manages your cleaning supplies with room to spare. Other features include a fold-away cart extension for easy storage and heavy-duty 4-inch casters for enhanced mobility.
Each mopping system component plays an important role in delivering the highest level of cleanliness. For example, our innovative swivel mop frame is accurately balanced to maintain total surface contact at any angle. The unique wringing mechanism exerts force evenly across the entire mop surface, extracting the maximum amount of soiled contaminants.
Specifications
Specifications
| Color | White |
| Dimensions (L x W x H) | 40 x 19 x 23 in. |