Learn More
Invitrogen™ TrueTag™ Donor DNA Kit, BFP
Description
- A rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments
- Kit provides everything needed to produce a high-quality DNA donor to tag the gene with a selection marker and BFP
- Simply provide two locus-specific primers to add homology arms, PCR-amplify, and purify for a transfection-ready donor in hours, not days
- One-step PCR amplification to generate donor templates – no cloning required
- Enrich up to 100% BFP-tagged cells
Each kit is designed to enable users to:
- Tag the carboxy- (C-) or amino- (N-) terminus of the gene of interest
- Choose from puromycin or blasticidin selection markers – whatever best suits the cell model
- Easily remove antibiotic resistance markers with Cre/Lox following cell enrichment
Included in the kit:
- Four linear donor templates: N-terminus/puromycin resistance, N-terminus/blasticidin resistance, C-terminus/puromycin resistance, C-terminus/blasticidin resistance
- Phusion Flash High-Fidelity PCR Master Mix
- PCR cleanup columns and buffers
- Positive control primers to tag human beta-actin (ACTB)
Intuitive software to rapidly design gene-specific reagents
Target-specific gene editing reagents are required for proper knock-in of your fluorescent tag. The Invitrogen TrueDesign Genome Editor web-based software does the design work for you:
- Identifies target gene transcript
- Designates insertion site
- Designs and enables easy ordering of all necessary components
Required reagents available via TrueDesign software (not included in kit):
- CRISPR guide RNA or TALENs designed adjacent to N- or C-terminus
- Oligos to PCR-amplify the donor and add appropriate homology arms
- Sequencing primers for assessment of CRISPR-Cas9 cleavage efficiency
- Verification primers to validate proper insertion of the TrueTag DNA Donor
Also required and available for purchase separately or as part of TrueDesign workflow:
- TrueCut Cas9 Protein v2, TrueCut HiFi Cas9 Protein, or GeneART CRISPR mRNA
- CRISPRMAX Transfection Reagent or MessengerMAX Transfection Reagent
Specifications
Specifications
| For Use With (Application) | Cellular Imaging |
| No. of Reactions | 10 reactions |
| Quantity | 10 Reactions |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.