Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trx-Tag Monoclonal Antibody (7G10), HRP, Bioss
SDP

Catalog No. 502280023
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0023 Supplier Bioss Supplier No. BSM33018MHRP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Trx-Tag Monoclonal antibody specifically detects Trx-Tag in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Trx-Tag
Applications Western Blot
Classification Monoclonal
Clone 7G10
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene TXNRD3
Gene Accession No. Q86VQ6, Q99MD6
Gene Alias thioredoxin
Host Species Mouse
Immunogen Recombinant human Thioredoxin (amino acids 1-105).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114112, 232223, 297437
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.