Learn More
Gibco™ TrypLE™ Express Enzyme (1X), no phenol red

Description
Cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells.
- Gentle on cells
- Room temperature-stable for 24 months
- Easy to use
- Animal origin-free
- Gentle on cells, purity increases specificity due to the action of a single enzyme and reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts
- Can be directly substituted for trypsin in existing protocols
- Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS
Energy efficient, less hazardous, sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP-compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Chelators | EDTA |
| Content And Storage | Storage conditions: 15°C to 30°C, protect from light Alternate storage conditions: 2°C to 8°C or -5°C to -20°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
| Cell Type | Mammalian Cells |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Culture Type | Adherent Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Dissociation Reagent |
| Sterility | Sterile-filtered |
| Without Additives | No Phenol Red |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.