Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Trypsin (Bovine), Irradiated

Catalog No. 89965894 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-894 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-965-894 Supplier Abnova™ Supplier No. P5259
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number Q29463
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282603/615237
Name Trypsin (Bovine), Irradiated
Preparation Method Native protein chromatographically purified from Bovine Pancreas
Quantity 100 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Gene Alias TRYP8
Common Name PRSS2
Gene Symbol PRSS2
Biological Activity ≥180 TAME units per mg protein. TAME Unit: One Unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 10mM calcium.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Bovine Pancreas
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.