Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Trypsin (Bovine), Sequencing Grade

Catalog No. 89965889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
4 x 25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-889 4 x 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-965-889 Supplier Abnova™ Supplier No. P5319
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number Q29463
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 282603/615237
Name Trypsin (Bovine), Sequencing Grade
Preparation Method Native protein purified from Bovine Pancreas
Quantity 4 x 25 μg
Storage Requirements Store at -20°C on dry atmosphere.
Regulatory Status RUO
Gene Alias TRYP8
Common Name PRSS2
Gene Symbol PRSS2
Biological Activity ≥150 TAME units per mg protein (at least 8,625 BAEE/2875 USP/NF units per mg protein). TAME Unit: One Unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 10mM calcium.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Pancreas
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.