Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Trypsin Inhbitor (Ovomucoid) (Egg White)

Catalog No. 89965896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-896 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-965-896 Supplier Abnova™ Supplier No. P5243
Add to Cart
Add to Cart

Used for IA

#REF!

Specifications

For Use With (Application) Immunoassay
Molecular Weight (g/mol) ∽28kDa
Name Trypsin Inhibitor (Ovomucoid) (Egg White)
Preparation Method Mucoprotein and antitryptic factor purified from egg white described by Lineweaver and Murray, J. Biol. Chem., 171, 565 (1947)
Quantity 1 g
Storage Requirements Store at 4°C. Reconstituted with 0.001 N HCl at a concentration of 1.0 mg/ml.
Regulatory Status RUO
Common Name TRYPSIN
Biological Activity 1mg inhibits at least 1.2 mg trypsin (Cat. # P5255).
Species Chicken
Protein Tag None
Expression System Mucoprotein and antitryptic factor purified from egg white described by Lineweaver and Murray, J. Biol. Chem., 171, 565 (1947)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.