Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Trypsin Inhibitor, chicken egg whites

Catalog No. AAJ63927ME
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63927ME 500 mg
AAJ6392703 1 g
AAJ6392704 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63927ME Supplier Thermo Scientific Chemicals Supplier No. J63927ME
Get 15% Off Thermo Scientific Reagents
Use promo code REAGENTS15
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits the pancreatic enzyme trypsin | CAS: 9035-81-8

Soybean trypsin inhibior (STI) is a protein which reduces trypsin (a proteolytic enzyme) as well as plasma kallikrein, factor Xa and plasmin activity. Soybean trypsin inhibitor has demonstrated inhibition of human neutrophil elastase at an IC50=0.3 nM. At this concentration soybean trypsin inhibitor showed neither cytotoxic nor haemolytic effects on populations of human blood cells. In fibrin matrix, and without toxic effects, STI had a strong inhibitory effect on human umbilical vein endothelial cells migration and tubulogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9035-81-8
Synonym Trypsin inhibitor, ovomucoid

Specifications

Quantity 500 mg
Description White
For Use With (Application) Inhibits the pancreatic enzyme trypsin
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.