Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Trypsin Inhibitor III, 1 mg/ml in PBS, animal and human components free, 250 ml (20 x 50 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P10034100X20
Encompass_Preferred
PAN-Biotech Trypsin Inhibitor III: 1 mg/ml in PBS, free of animal & human components. QUANTITY: 20 x 50 ml. STORAGE: -20 degrees Celsius. MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: Trypsin Inhibitor III is a sterile solution of purified natural protease inhibitor proteins proteins to inhibit trypsin proteolytic activity. It does not contain any animal or human components. FORMULATION: Supplied as 1 mg/ml in PBS solution. APPLICATION: For inhibiting trypsin activity in serum-free culture of adherent cells. Trypsin is used for dissociation of adhesion-dependent cells. Serum contains natural trypsin inhibitors. In serum-free cell culture the neutralizing effect of serum is absent. Therefore, it is crucial to completely block proteolytic activity from trypsin after detachment of the cells. Trypsin Inhibitor III is cell culture tested and is appropriate for use in cell culture applications.
Catalog No. 50-238-8472
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.