Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Trypsin Monoclonal Antibody (TRYP11-236.7), Invitrogen™

Catalog No. PIMA546066
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546066 100 μg
01-671-570 500 μg
PIMA546011 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546066 Supplier Invitrogen™ Supplier No. MA546066
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10617 detects Trypsin from human samples. MA1-10617 has been successfully used in ELISA applications. The MA1-10617 immunogen is purified human pancreatic Trypsin.

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin
Applications ELISA
Classification Monoclonal
Clone TRYP11-236.7
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PRSS1
Gene Accession No. P07477
Gene Alias Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsin I; anionic trypsin-1; Anionic trypsinogen; beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pretrypsinogen I; Protease Serine 1; protease serine 2 preproprotein; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine 2; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS1; PRSS2; PTRYI; RATPTRYI; Serine protease 1; Serine protease 2; TCR V beta 4.1; TRP1; Try1; Try-1; TRY2; TRY4; TRY8; Trygn16; TRYP1; TRYP2; TRYP8; trypsin 1; Trypsin I; Trypsin II; trypsin-1; Trypsin-2; trypsinogen; trypsinogen 1; trypsinogen 16; trypsinogen 2; trypsinogen A
Gene Symbols PRSS1
Host Species Mouse
Immunogen Purified human pancreatic Trypsin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5644
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.