Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation Trypsin Platinum, Mass Spec Grade

Catalog No. PRVA9000 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRVA9000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRVA9000 Supplier Promega Corporation Supplier No. VA9000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Achieve superior autoproteolytic resistance with Recombinant Trypsin for most accurate biotherapeutic protein characterization and specialized proteomics applications. Free of chymotryptic activity, works with existing trypsin protocols, ensuring excellent lot-to-lot reproducibility and free of contaminating animal proteins.

  • Recombinant trypsin for accurate biotherapeutic protein characterization.
  • Free of chymotryptic activity.
  • Superior autoproteolytic resistance.
  • Works with any existing trypsin protocols.
  • Excellent lot-to-lot reproducibility.
  • Free of contaminating animal proteins
  • Suitable for specialized proteomics applications
  • High-quality trypsin for consistent performance
  • Ideal for mass spectrometry and other analytical techniques
  • Reliable and reproducible results
Storage Conditions:  Store lyophilized Trypsin Platinum, Mass Spectrometry Grade, at –30°C to –10°C. Reconstitute the enzyme in 50mM acetic acid, dispense into aliquots and store at –30°C to –10°C for up to 1 month. For longer storage, freeze the reconstituted Trypsin Platinum solution at –80°C to –60°C for up to 6 months. Reconstituted Trypsin Platinum solution retains full activity after three freeze-thaw cycles.

Specifications

For Use With (Equipment) Mass Spectrometer
Solubility Soluable in digestion buffers
Product Type Protease
Content And Storage Store lyophilized Trypsin Platinum, Mass Spec Grade, at –30°C to –10°C.
Starting Material Protein Samples
Final Product Type Digested Peptides
Detection Method Fluorescence
Format Bottle
Workflow Step Protein digestion
Quantity 100 μg
For Use With (Application) Protein digestion for mass spectrometry analysis
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.