Description
- Tryptic Soy Broth (TSB) or Casein Soya Bean Digest Broth is a highly nutritious medium used for the cultivation of a wide range of microorganisms
- It is recommended by the harmonized USP/EP/JP method for sterility testing and for microbiological examination of non-sterile products
- TSB supplemented with 15% glycerol (v/v) is used to maintain bacterial viability for extended periods by freezing
- Liquid medium in plastic tubes
Specifications
Specifications
| Manufacturing Origin | Italy |
| Product Type | Broth Tube |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Cultivation |