Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptic Soy Broth
GSA_VA

Catalog No. R064890
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
16mm x 125mm Tube
16mm x 125mm Tube, Hungate Cap
20mm x 113mm Tube
20mm x 150mm Tube
D - 1mL
D - 2mL
Tube
Fill Volume:
1 mL
10 mL
10 mL HC
15 mL
2 mL
20 mL
4.5 mL
5 mL
9 mL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Fill Volume
R064890 100/Pk. D - 2mL 2 mL
R065000 100/Pk. 15mm x 103mm Tube 4.5 mL
R07222 100/Pk. Tube 5 mL
R07226 100/Pk. Tube 10 mL
R117832 100/Pk. 16mm x 125mm Tube 9 mL
R117834 20/Pk. 16mm x 125mm Tube, Hungate Cap 10 mL HC
R118881 20/Pk. 20mm x 113mm Tube 15 mL
R119880 100/Pk. 20mm x 150mm Tube 20 mL
R064880 100/Pk. D - 1mL 1 mL
R07224 20/Pk. Tube 5 mL
R08942 100/Pk. 20mm x 113mm Tube 10 mL
R064892 20/Pk. D - 2mL 2 mL
R07228 20/Pk. Tube 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. R064890 Supplier Thermo Scientific™ Supplier No. R064890
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.

Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.

  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Tryptic Soy Broth
Format D - 2mL
Manufacturing Origin United States
Product Type Microbiology Media
Fill Volume 2 mL
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.