Learn More
Description
- Tryptic Soy Broth is widely used for the isolation of bacteria from clinical specimens, supporting the growth of a majority of pathogenic bacteria
- This medium contains digests of soybean meal and casein, which provide amino acids and other nitrogenous substances, making it a highly nutritious medium for a variety of organisms, including most pathogenic bacteria
- Dipotassium phosphate is included in the formulation as a buffer to maintain the pH
Specifications
Specifications
| Appearance | Clear |
| ATCC® | 16404, 10231, 6633, 9027, 6538 |
| Broth Type | Tryptic Soy Broth |
| Certifications/Compliance | USP |
| Color | Medium Amber |
| Content And Storage | 2°C to 30°C |
| Country of Origin | United States |
| Detects | Bacteria, Fungi, Yeast |
| Form | Liquid |
| Format | Bottle |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
