Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Tryptic Soy Broth (TSB), Polycarbonate Bottle
SDP

Catalog No. 23001773 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-773 500 mL
23-001-782 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-773 Supplier Hardy Diagnostics™ Supplier No. U65
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi

  • Heavy duty:Bottles are tough and sturdy to reduce breakage
  • Optically clear:Bottles are made of clarified polycarbonate for clear viewing
  • Easy to read:Each bottle is clearly marked with a measuring fill line

Specifications

Appearance Clear
ATCC® 6538, 9027, 6633, 10231, 16404
Broth Type Tryptic Soy Broth
Certifications/Compliance ISO 13485, ISO 17025, ANAB Calibration ISO 17025, BSI-ISO 13485
Color Light Amber
Content And Storage 20°C to 25°C
Country of Origin United States
Additive Sodium Chloride, Dextrose, Dipotassium Phosphate
Detects Bacteria, Yeast
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Tryptic Soy Broth
Product Type Broth
Target Organism Class Non-fastidious Organisms
Test Type Microbial Cultivation
Fill Volume 500 mL
Final Product Type Bottle and Jar Media (Broth)
Formulation Ingredients per liter of deionized water-Pancreatic Digest of Casein 17.0gm, Sodium Chloride 5.0gm, Papaic Digest of Soybean Meal 3.0gm, Dextrose 2.5gm, Dipotassium Phosphate 2.5gm
For Use With (Application) Cultivation, Colony Counts
Incubation Time 18 to 24 hr.
Material Polycarbonate
Packaging Type Polycarbonate Bottle
Quantity 500 mL
Shipping Condition Room Temperature
Specimen Type Environmental and Industrial Sources
Protein Source Casein
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.