Learn More
Teknova Tryptic Soy Broth with 0.1% Tween-80, 1000mL, Sterile. 8 pack.
SDP
Supplier:  Teknova T15748PK
Encompass_Preferred
1.70% Tryptone, 0.30% Soytone, 0.25% Cerelose Anhydrous Dextrose (Glucose), 0.50% Sodium Chloride,0.25% Potassium Phosphate Dibasic, 0.10% Tween-80
Catalog No. 50-238-7437
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.