Learn More
Teknova Trypticase Soy Agar Plate. 150mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova T51444PK
Encompass_Preferred
1.50% Tryptone, 0.50% Soytone, 0.50% Sodium Chloride, 1.50% Agar
Catalog No. 50-238-7724
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.