Learn More
United States Biological Corporation Tryptone (BSE-Free) | 500g
Catalog No. NC1773008 Shop All United States Biological Corporation Products
Encompass_Preferred
Tryptone is a specially formulated enzymatic digest of casein, similar to Peptone from gelatin. It is used to supply higher growth rates when used in molecular biology media preparations such as YPD Broth and Agar. Used as a nitrogen source in culture media for fastidious and non-fastidious organisms. Also used to differentially select bacteria based on carbohydrate intake since Tryptone is carbohydrate deficient.
Catalog No. NC1773008 Supplier United States Biological Corporation Supplier No. T8750500G
Tryptone is a specially formulated enzymatic digest of casein, similar to Peptone from gelatin. It is used to supply higher growth rates when used in molecular biology media preparations such as YPD Broth and Agar. Used as a nitrogen source in culture media for fastidious and non-fastidious organisms. Also used to differentially select bacteria based on carbohydrate intake since Tryptone is carbohydrate deficient.