Learn More
Description
Grow a wide variety of microorganisms with Thermo Scientific ™ Oxoid ™ Tryptone Soya Agar (Dehydrated). A general purpose agar medium containing two peptones, Tryptone Soya Agar supports the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition1.
Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopeia and food testing methods for the growth of a wide variety of organisms. Recommended as a reference medium for testing selective media. Conforms to formulations detailed in the international pharmacopoeia2,3,4,5.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Tryptone Soya Agar |
| Form | Powder |
| Product Type | Tryptone Soya Agar |
| Quantity | 2.5 kg |
| Yield | For 62.5L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.