Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophan hydroxylase 2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB338027
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338027 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338027 Supplier R&D Systems Supplier No. NBP177556APC

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Tryptophan hydroxylase 2
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [Uniprot: Q8IWU9]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 121278
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.