Learn More
Description
Cultivate and isolate fastidious microorganisms and determine their hemolytic reactions using Thermo Scientific™ Oxoid™ Tryptose Blood Agar Base (Dehydrated) with supplementation of blood. This highly nutritious medium is specially developed by Casman 1,2 for the preparation of a blood agar which will support the growth of many fastidious organisms.
The original formulation included 0.3g dextrose per litre which interfered with the hemolytic reactions. It is now used without dextrose as a standard medium3.
Tryptose Blood Agar Base with added blood gives good hemolytic reactions and without blood, it will sustain good to excellent growth of many demanding organisms. However, to improve the growth of certain organisms e.g. Neisseria meningitidis and Streptococcus pneumoniae, the addition of 1g of yeast extract (LP0021) to one liter of Tryptose Blood Agar Base is recommended.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Tryptose Blood Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 16.7L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.