Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptose Phosphate Broth (Dehydrated)

Catalog No. OXCM0283B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0283B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0283B Supplier Thermo Scientific™ Supplier No. CM0283B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate fastidious bacteria and supplement cell culture media with this medium.

Cultivate fastidious bacteria with Thermo Scientific™ Oxoid™ Tryptose Phosphate Broth (Dehydrated). Tryptose Phosphate Broth is recommended for the cultivation of streptococci, pneumococci, meningococci and other fastidious organisms. Tryptose Phosphate Broth with added agar and sodium azide is recommended for the isolation of pathogenic streptococci from cheese and other dairy products1,2.

Use Tryptone Phosphate Broth for cultivation of fastidious bacteria.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: FDA.
Description Tryptose Phosphate Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 16.9L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.