Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSEN15 Monoclonal Antibody (OTI5A3), TrueMAB™, OriGene
SDP

Catalog No. 501684660
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4660 Supplier OriGene Supplier No. TA812717
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Non-catalytic subunit of the tRNA-splicing endonuclease complex, a complex responsible for identification and cleavage of the splice sites in pre-tRNA. It cleaves pre-tRNA at the 5′ and 3′ splice sites to release the intron. The products are an intron and two tRNA half-molecules bearing 2′,3′ cyclic phosphate and 5′-OH termini. There are no conserved sequences at the splice sites, but the intron is invariably located at the same site in the gene, placing the splice sites an invariant distance from the constant structural features of the tRNA body. The tRNA splicing endonuclease is also involved in mRNA processing via its association with pre-mRNA 3′-end processing factors, establishing a link between pre-tRNA splicing and pre-mRNA 3′-end formation, suggesting that the endonuclease subunits function in multiple RNA-processing events.

Specifications

Antigen TSEN15
Applications Western Blot
Classification Monoclonal
Clone OTI5A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TSEN15
Gene Accession No. Q8WW01
Gene Alias C1orf19, PCH2F, sen15
Gene Symbols TSEN15
Host Species Mouse
Immunogen Full length human recombit protein of human TSEN15 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116461, 289083
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.