Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSG-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728652 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8652 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8652 Supplier Proteintech Group Inc Supplier No. 133211AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSG6 is a secretory protein that contains a hyaluro-binding domain, and thus is a member of the hyaluro-binding protein family. The hyaluro-binding domain is involved in extracellular matrix stability and cell migration. This protein forms a stable complex with inter-alpha-inhibitor (I alpha I), and thus enhances the serine protease inhibitory activity of I alpha I, which is important in the protease network associated with inflammation. The expression of this gene can be induced by tumor necrosis factor alpha and interleukin-1. The expression can also be induced by mechanical stimuli in vascular smooth muscle cells, and is correlated with proteoglycan synthesis and aggregation.

Specifications

Antigen TSG-6
Applications Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TNFAIP6
Gene Accession No. O08859, P98066
Gene Alias Hyaluronate binding protein, TNF alpha induced protein 6, TNF stimulated gene 6 protein, TNFAIP6, TSG 6, TSG6
Gene Symbols TNFAIP6
Host Species Rabbit
Immunogen TSG-6 Fusion Protein Ag3885
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21930, 7130, 84397
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.