Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSG Series General Purpose Laboratory Freezers

Catalog No. TSG25FSSA
Change view
Click to view available options
Capacity (English):
25 cu. ft.
49 cu. ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
TSG25FSSA 25 cu. ft.
TSG49FSSA 49 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSG25FSSA Supplier Thermo Scientific™ Supplier No. TSG25FSSA
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Thermo Scientific™ TSG Series general purpose freezers are designed to meet the general storage needs of research, clinical and industrial laboratories.

The Thermo Scientific™ TSG Series general purpose laboratory auto-defrost freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

Utilizing natural refrigerants in accordance with the U.S. Environmental Protection Agency (EPA) Significant New Alternative Policy (SNAP) and energy efficient systems that enable ENERGY STAR certification, the TSG Series improves scientific and healthcare sustainability.

The TSG Series general purpose laboratory freezers provide a reliable, quiet and efficient work environment.

Enabling lab efficiency and compliance

  • Designed to safely store reagents, media, proteins, and samples used daily in the lab which require great performance
  • Adaptive defrost with time and temperature guided cycle provides safe storage temperatures and allows for continuous use without the need for defrost
  • Easy to use digital controller with setpoint security and alarms (temperature, door ajar, and power failure)
  • Tested and certified to conform with the design and safety standards of UL 471

Designed for an ergonomic and comfortable experience

  • Easy to clean stainless steel surfaces and epoxy coated shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the freezer without distraction
  • Self-closing door with 90° stop to assist with inventory loads
  • Standard door locks provide comfort and assurance

Verified sustainability

  • US EPA ENERGY STAR Certified
  • Natural refrigerants with near 0 GWP and ODP make the TSG Series compliant with US EPA's Significant New Alternatives Policy (SNAP)

Ready for monitoring

  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
  • Built in remote alarm terminals

Services and support

  • 24 Months parts and labor, 60-month compressor parts warranty (varies by country, please contact us for details)
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 25 cu. ft.
Capacity (Metric) 700 L
Temperature Range -12°C to -30°C
Temperature (Setpoint) -23C°
Door Style Solid
No. of Doors 1
Door Material Stainless Steel
Cabinet Material Stainless Steel
Interior Stainless Steel
Shelves 4
Capacity (English) Shelf 100 lb.
Capacity (Metric) Shelf 45 kg
Certifications/Compliance ENERGYSTAR, Prop 65, cULus
Casters Yes
Control Type Digital Micrprocessor
Ports 1 Rear Access Port
Warranty 2 years parts and labor, plus 3-year compressor warranty
Defrost Automatic
Dimensions (DxWxH) Exterior 27.75 x 32 x 81 in. (705 x 813 x 2057mm)
Dimensions (D x W x H) Interior 24 x 28.5 x 61.75in. (610 x 724 x 1569mm)
Dimensions (L x W x H) Exterior 27.75 x 32 x 81in. (705 x 813 x 2057mm)
Access Security Key Lock
Amperage 8.8A
Plug Type NEMA 5-15 P
Electrical Requirements 115V 60Hz
Frequency 60 Hz
Refrigerant R290
Shipping Weight (English) 392 lb.
Shipping Weight (Metric) 178kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.