Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSH beta Mouse anti-Human, Clone: 2G1H3, Proteintech
SDP

Catalog No. 501737158 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7158 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7158 Supplier Proteintech Group Inc Supplier No. 667501IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.

Specifications

Antigen TSH beta
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2G1H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TSHB
Gene Accession No. P01222
Gene Alias CHNG4, Thyrotropin alfa, Thyrotropin beta chain, Thyrotropin subunit beta, TSH B, TSH BETA, TSHB
Gene Symbols TSHB
Host Species Mouse
Immunogen TSH beta Fusion Protein Ag27153
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7252
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.