Learn More
BioVendor, LLC Thyroid Stimulating Hormone (TSH) canine ELISA, 96 wells
SDP
Supplier:  BioVendor, LLC RTC019R
Encompass
Thyroid Stimulating Hormone (TSH) canine ELISA. Research use only. Immunoassay kit for quantification of canine TSH in serum or plasma samples. Calibration range: 0.25.2 ng/ml. Detection limit: 0.01 ng/ml. Synonyms: thyrotropin, thyrotropic hormone. Summary: TSH in dogs is similar in function to TSH in other mammals, including humans. Acting on the thyroid gland, it plays a major role in maintaining normal levels of iodothyronines, T4 and T3. Production and secretion of TSH is controlled by negative feedback from circulating T4 and T3, and by TRH (thyrotropin releasing hormone). Hypothyroidism is common in dogs, whereas hyperthyroidism in dogs is nearly unknown. Usually, hypothyroidism in dogs is suspected on basis of clinical history and presence of lowered levels of thyroid hormones. However, lowered thyroid hormone levels are nonspecific indicators, as they are also observed in nonthyroid illnesses. Determination of canine TSH may improve evaluation of thyroid function in dogs.
Catalog No. 50-206-9300
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.