Learn More
Drg International Inc Tsh Elisa Ruo
Supplier:  Drg International Inc EIA4171R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of TSH (Thyroid Stimulating Hormone) in serum or plasma. This product is for Research Use Only. The DRG TSH ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site of the TSH molecule.
Catalog No. 50-194-7606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.