Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSH Receptor Monoclonal Antibody (2C11)
GREENER_CHOICE

Catalog No. PIMA528136 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528136 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528136 Supplier Invitrogen™ Supplier No. MA528136
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

TSH Receptor Monoclonal Antibody for Western Blot, Flow, ELISA, IP

TSH Receptor gene is a membrane protein and a major controller of thyroid cell metabolism. The encoded protein is a receptor for thyrothropin and thyrostimulin, and its activity is mediated by adenylate cyclase. Defects in this gene are a cause of several types of hyperthyroidism. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSH Receptor
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Western Blot
Classification Monoclonal
Clone 2C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene TSHR
Gene Accession No. P16473
Gene Alias AI481368; CHNG1; hTSHR-I; hypothroid; hyt; LGR3; MGC141920; pet; seven transmembrane helix receptor; thyroid stimulating hormone receptor; thyroid stimulating hormone receptor, isoform 2; thyroid-stimulating hormone receptor; thyroid-stimulating hormone receptor variant; thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSHR; TSH-R; TSHR; seven transmembrane domain G protein-coupled receptor; TSHRA
Gene Symbols TSHR
Host Species Mouse
Immunogen Recombinant Human TSH receptor.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.