Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHZ1 Monoclonal Antibody (OTI8A9), TrueMAB™, OriGene
SDP

Catalog No. p-200015632
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3142 Supplier OriGene Supplier No. TA809993
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. TSHZ1 has been found to be required for axial skeleton, soft palate and middle ear development in mice and may be involved in a common pathway with the Hox genes. Both TSHZ1 and the related protein TSHZ3 have been found to interact with FE65, an adapter protein that binds to the amyloid protein precursor (APP) in neurons. Together with SET, a component of the inhibitor of acetyl transferase, and histone deacetylases, these proteins formed a gene-silencing complex whose target includes caspase-4.

Specifications

Antigen TSHZ1
Applications Western Blot
Classification Monoclonal
Clone OTI8A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TSHZ1
Gene Accession No. Q6ZSZ6
Gene Alias CAA, NY-CO-33, SDCCAG33, TSH1
Gene Symbols TSHZ1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 29-176 of human TSHZ1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10194
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.