Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHZ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103361
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103361 100 μL
1 options

Catalog No. PIPA5103361

Supplier: Thermo Scientific PA5103361

Only null left
Add to Cart
Add to Cart

TSHZ2 Polyclonal Antibody

TSHZ2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSHZ2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSHZ2
Gene Accession No. Q68FE9, Q9NRE2
Gene Alias 2900073F20Rik; B830045G17; C20orf17; cell growth-inhibiting protein 7; Kiaa4248; mKIAA4248; Mtsh2; Ovarian cancer-related protein 10-2; OVC10-2; Sdccag33l; SDCCAG33-like protein; serologically defined colon cancer antigen 33 like; teashirt 2; teashirt family zinc finger 2; teashirt homolog 2; teashirt homolog 2-like; teashirt zinc finger family member 2; teashirt zinc finger homeobox 2; teashirt2; TSH2; TSHZ2; ZABC2; Zfp218; Zinc finger protein 218; ZNF218
Gene Symbols TSHZ2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSHZ2(Accession Q9NRE2), corresponding to amino acid residues D225-L275.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128553, 228911
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.