Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propan-2-yl 4-(3,4-dichloro-2,5-dioxopyrrol-1-yl)benzoate | 704878-75-1 | 98.5% | 328.15 g/mol | C14H11Cl2NO4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1128255MG

Encompass_Preferred

TSI-01 is a research-grade small molecule inhibitor targeting lysophosphatidylcholine acyltransferase 2 (LPCAT2). It is supplied as a 5 mg solid for laboratory use and is intended for in vitro biochemical and cell-based assays probing LPCAT2-mediated lipid remodeling and inflammatory signaling. CAS number: 704878-75-1.

  • Selective LPCAT2 inhibitor (IC50 = 0.47 μM for human LPCAT2).
  • High purity suitable for research use (98.52%).
  • Molecular formula C14H11Cl2NO4; molecular weight 328.15 g/mol.
  • Supplied as a 5 mg solid, convenient for small-scale studies.
  • Soluble in DMSO for assay preparation.

Catalog No. 50-002-62991


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.