Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST Q 10X100
Supplier:  Tosoh Bioscience LLC 0021410
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel BioAssist Q; Matrix: polymethacrylate; Material: PEEK; Counter ion: Clsup- Particle size: 13u m; Pore size: 4000angstrom pH: 2.0 -12; 10mm I.D. x 10cm length
Catalog No. 50-851-674
Only null left
Add to Cart
Request a Quote