Learn More
Tosoh Bioscience LLC TSKGEL GMHXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0016141
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel GMHXL; Media: Mixed bed polymer; Particle: 9 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-400