Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ HHR Guard Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021899 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
4 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-1899 4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1899 Supplier MilliporeSigma Supelco Supplier No. 817368
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a broad range of organic solvents.

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 4 cm
Pore Size Mixed
For Use With (Equipment) G1000HHR to G4000HHR and GMHHR-L Columns
Temperature (Metric) Maximum 80°C
Packing Material Polymer Particle Platform
Technique Size Exclusion (SEC)
USP Type L21

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.