Learn More
TSKgel™ Hardware and Accessories - Holder, Sold by MilliporeSigma™ Supelco™
Catalog No. 111021931 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1/Pk.
Catalog No. Quantity
11-102-1931 1/Pk.
1 options
Catalog No. 11-102-1931 Supplier MilliporeSigma Supelco Supplier No. 818206
Add to Cart
Add to Cart
Holder for TSKgel™ Super Series Guardfilters
  • Protect your 4.6 mm ID TSKgel Super Series column (Super-Octyl, Super-ODS and Super-Phenyl) from plugging by installing a guard filter in front of the analytical column.
  • The Holder for Super Series Guardfilters requires 0.4 cm L x 4 mm ID Guardfilters (PN 818207) that need to be purchased separately.
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC: Suitable, L1 Per USP
Quantity 1/Pk.
Technique Reverse Phase HPLC
TSKgel is a registered trademark of Tosoh Corporation