Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Octadecyl-2PW Guard Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022212 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Pore Size: 125 Å
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2212 Pore Size: 125 Å
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2212 Supplier MilliporeSigma Supelco Supplier No. 842161
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guard cartridge

  • Highly cross-linked polymethacrylate base material of TSKgel Octadecyl-2PW provides excellent stability in high pH buffer systems and can withstand rigorous cleaning with either acid or base.
  • The 125Ã… pores allow for anaylsis of peptides up to 8,000 Da.
  • Columns packed with larger particle sizes are available for semi-preparative scale separations.
  • TSKgel Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No.819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No.819018.
  • Reversed-phase liquid chromatography (RPLC) is the preferred method for analysis and purification of polar and nonpolar compounds that are soluble in mixtures of water and organic solvents.
  • 0-100% organic solvent
TRUSTED_SUSTAINABILITY

Specifications

Quantity 3/Pk.
pH 2 to 12
Particle Size 5 μm
Length (Metric) 1 cm
Pore Size 125 Å
Diameter (Metric) Inner 2 mm
For Use With (Equipment) HPLC: Suitable
Stationary Phase C18 (octadecyl)
Temperature (Metric) Maximum 4°C to 50°C
Technique Reversed phase
Matrix Polymer particle platform matrix
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.