Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Octadecyl-2PW Guard Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021907 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
1 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-1907 1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1907 Supplier MilliporeSigma Supelco Supplier No. 817503
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel Guard Columns are stand-alone and do not require separate holders. For use with 6 mm ID TSKgel Octadecyl-2PW Columns.

  • Shipped in 55% Acetonitrile, 45% Water
TRUSTED_SUSTAINABILITY

Specifications

pH 2 to 12
Particle Size 5 μm
Length (Metric) 1 cm
Pore Size 125 Å
Diameter (Metric) Inner 6 mm
Description 1 meq/mL (Estimated Ligand Density)
Stationary Phase C18 (octadecyl)
Temperature (Metric) Maximum 50°C
Solvents 0-100% Organic Solvent
Packing Material Polymer Particle Platform
Phase Reversed Phase
USP Type L1
Product Line TSKgel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.