Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 2X50 3UM
Supplier:  Tosoh Bioscience LLC 0021812
Encompass_Preferred
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 3 micrometer; Pore Size: 100 angstrom;2.0mm I.D. x 5cm Length; Stainless Steel
Catalog No. 50-851-698
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.