Learn More
Tosoh Bioscience LLC TSKGEL ODS-100Z 4.6X250 5UM
Supplier:  Tosoh Bioscience LLC 0021462
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100Z; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;4.6mm x 25cm Length; Stainless Steel
Catalog No. 50-851-687