Learn More
Tosoh Bioscience LLC TSKgel Protein A-5PW | 20um | PEEK | 4.6mm ID | 3.5cm Length
Supplier:  Tosoh Bioscience LLC 0023483
Encompass
The TSKgel Protein A-5PW column has been designed for the rapid separation and robust quantification of a variety of antibodies. Monoclonal antibodies from harvested cell culture media can be captured and accurately quantitated in less than 2 minutes per injection. The column can be used for more than 2,000 injections without regeneration or cleaning. Packed with hydroxylated methacrylic polymer beads, the TSKgel Protein A-5PW column is designed with a high degree of crosslinking, which allows high flow rate for chromatography while still maintaining chromatographic efficiency, peak width and resolution. The recombinant protein A ligand is a code-modified hexamer of the C domain. An enhanced rProtein A ligand is bound to the TSKgel 5PW base bead via multipoint attachment resulting in excellent base stability in 0.1 mol/L NaOH.
Catalog No. NC1304454
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.