Learn More
Tosoh Bioscience LLC TSKGEL SUPER-OCTYL 4.6X50
Supplier:  Tosoh Bioscience LLC 0018275
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Super-Octyl; Media: Silica; Particle: 2 micrometer; Pore Size: 110 angstrom; 4.6mm I.D. x 5cm Length; Super Series
Catalog No. 50-851-486