Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Super-Phenyl Guard Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022215 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
PH: 2 - 7.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2215 PH: 2 - 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2215 Supplier MilliporeSigma Supelco Supplier No. 842177
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guard cartridge

  • Requires Guard Cartridge Holder, PN 819308
  • Packed with monodispersed 2 micron (nominal, mean particle size is 2.3 micron) spherical silica particles covalently bonded with phenyl groups.
  • The small particle size used in Super Series and TSKgel ODS-140HTP columns provides the highest efficiency of all TSKgel reversed phase columns.
  • The monodispersity of the particles reduces column back pressure to values more typical of larger particles, thus allowing the use of higher flow rates than other 2 micron packings.
  • TSKgel Super-Phenyl columns are less hydrophobic than TSKgel Super-Octyl and TSKgel Super-ODS columns.
  • They are an excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules and food/beverage samples.
  • 3% Carbon, multilayer, endcapped
  • No limitation salt concentration
  • 0-100% organic solvent
TRUSTED_SUSTAINABILITY

Specifications

pH 2 - 7.5
Particle Size 2.3 μm
Length (Metric) 1 cm
Surface Area 100 m2/g
Diameter (Metric) Inner 2 mm
Stationary Phase Phenyl
Temperature (Metric) Maximum 10°C to 50°C
Technique Reversed phase
Matrix Spherical silica matrix

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.