Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SuperSW mAb HTP Guard Column, 4 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022198 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter: 3 mm; Length: 2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2198 Diameter: 3 mm; Length: 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2198 Supplier MilliporeSigma Supelco Supplier No. 822858
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ SuperSW mAb columns are designed for the separation and analysis of monoclonal antibodies (mAb).

TSKgel™ SuperSW mAb HTP columns provide high throughput analysis of mAb monomer and dimer and is UHPLC compatible. These short, narrow bore (15 cm x 4.6 mm I.D.) silica-based stainless steel columns offer reduced lot-to-lot variation, long column life, reduction of unspecified adsorption, and superior recovery of aggregates. The TSKgel™ SW mAb HTP columns utilize a unique pore-controlled technology, which produces a shallow calibration curve in the molecular weight region of a typical monoclonal antibody.

Order Info

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

TRUSTED_SUSTAINABILITY

Specifications

pH 2.5 - 7.5
Particle Size 4 μm
Length (Metric) 2 cm
Pore Size 250 Å
Diameter (Metric) 3 mm
Flow Rate 0.5 mL/min.
Stationary Phase Diol
Solvents 0 to 100% aqueous soluble organic solvents
Technique HPLC
USP Type L20, L59
Product Line TSKgel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.