Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ SWXL Top-Off Gel, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021811 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Bottle of 1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-1811 1 Bottle of 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1811 Supplier MilliporeSigma Supelco Supplier No. 808544
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diol phase matrix active group

  • Top-Off for TSKgel™ SWxl and QC-PAK columns, 5 μm, 1 gram TSKgel™ SWXL top-off gel may be used as a silica-based support column in the separation of proteins using high performance liquid chromatography (HPLC)
  • Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.
  • ≤0.5 M salt concentration
  • 0-100% aqueous soluble organic solvents
TRUSTED_SUSTAINABILITY

Specifications

For Use With L59 Per USP, L20 Per USP
Particle Size 5 μm
pH 2.5 - 7.5
Quantity 1 Bottle of 1 g
Temperature 10°C to 30°C
Matrix Silica particle platform

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.