Learn More
Tosoh Bioscience LLC TSKGEL TRESYL-5PW 6X40
Supplier:  Tosoh Bioscience LLC 0014455
Encompass_Preferred
Column, Affinity Chromatography; Tosoh TSKgel Tresyl-5PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 6mm I.D. x 4cm Length; Stainless Steel
Catalog No. 50-851-351
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.