Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ UP-SW2000 Guard Column for 4.6 mm ID Column, 2 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022206 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter: 4.6 mm; Length: 2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-102-2206 Diameter: 4.6 mm; Length: 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2206 Supplier MilliporeSigma Supelco Supplier No. 823516
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSKgel™ UP-SW2000 columns are designed for the analysis of small proteins, peptides and oligonucleotides.

TSKgel™ UP-SW2000 columns offer higher resolution, improved peak shape and increased efficiency yielding methods that are robust, reproducible and easily transferrable between UHPLC and HPLC systems. TSKgel™ UP-SW2000 columns offer superior reproducibility injection-to-injection, from column-to-column within the same lot and from lot-to-lot. %RSD values for peak parameters including retention times, area peak height, peak asymmetry, and theoretical plate count demonstrate the exceptional reproducibility of these columns.
TRUSTED_SUSTAINABILITY

Specifications

pH 2.5 - 7.5
Particle Size 2 μm
Length (Metric) 2 cm
Diameter (Metric) 4.6 mm
Material Stainless Steel Hardware
Technique HPLC
Product Line TSKgel

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.