Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TSLP ELISA Development Kit
SDP

Catalog No. DDXKETSLP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDXKETSLP 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDXKETSLP Supplier Novus Biologicals™ Supplier No. DDXKETSLP
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The human TSLP ELISA kit is for quantitative determination of TSLP concentrations in human cell culture supernatants and biological fluids (plasma, serum). This kit is for research purpose only. Kit contents: Capture Antibody: 0.5mg/mL of mouse anti-TSLP monoclonal antibody (clone 209A2.03). Dilute in carbonate buffer pH9.6 to a working concentration of 2.5μg/mL. Detection Antibody: 0.5mg/mL of Biotin-conjugated anti-TSLP mouse monoclonal antibody (clone210H1.04). Dilute to a working concentration of 3μg/mL in PBS-1% BSA-0.05% Tween20. Standard: Each vial contains 1ug/mL of recombinant TSLP produced, purified and concentrated from eukaryotic cells. A 7-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20 ng/mL is recommended. APPROPRIATE CONTROLS TO INCLUDE Following is a list of suggested controls to include with each analysis: 1. No capture antibody added to well 2. No lysate added to well 3. No capture antibody or lysate added to well 4. Positive control: use a cell line or tissue known to constitutively express p65 or a recombinantly expressed p65. 5. Negative control: use a cell line or tissue known to not express p65.

Specifications

Assay Sensitivity The minimum detectable dose of human TSLP was estimated at 1ng/mL
Product Type ELISA Kit
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 85480
Gene Symbol TSLP
Kit Contents Capture Antibody: 0.5mg/mL of mouse anti-TSLP monoclonal antibody (clone 209A2.03). Dilute in carbonate buffer pH9.6 to a working concentration of 2.5 ug /mL, Detection Antibody: 0.5mg/mL of Biotin-conjugated anti-TSLP mouse monoclonal antibody (clone 210H1.04). Dilute to a working concentration of 3ug/mL in PBS-BSA1%-Tween 0.05%, Standard: Each vial contains 1ug/mL of recombinant TSLP produced, purified and concentrated from eukaryotic cells. A 7-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20ng/mL is recommended
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym thymic stromal lymphopoietin
Target TSLP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.