Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSLP Monoclonal Antibody (eBio28F12 (28F12)), Functional Grade, eBioscience™, Invitrogen™

Catalog No. 16549185
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-549-185 500 μg
16-549-138 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-549-185 Supplier Invitrogen™ Supplier No. 16549185
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The MAb eBio28F12 reacts with and inhibits the bioactivity of mouse TSLP. Thymic Stromal-Derived Lymphopoetin (TSLP) was first identified in the tissue culture supernatant of a thymic stromal cell line as a growth factor capable of inducing proliferation and differentiation of pre-B cells. It was later found to be closely related to another stromal cytokine, IL-7, with which it shares overlapping functions. Along with its proliferative effects on B cells, TSLP induces dendritic cells to support the differentiation of naive T cells towards the Th2 lineage and may also be involved in the development of CD4^+CD25^+ regulatory T cells. The receptor for TSLP is heterodimeric and shares one subunit, IL-7Ralpha, with IL-7 and other members of the family, while the TSLPR subunit is unique to TSLP. TSLP expression can be induced by a variety of inflammatory cytokines and TLR ligands. Expression is regulated by NF-kappaB and has been detected in epithelial cells, stromal cells, and basophils. Elevated levels of TSLP are associated with asthma and atopic dermatitis, which are both Th2-mediated inflammatory conditions. It is believed that chronic overexpression of TSLP may result in increased sensitivity to allergens, resulting in susceptibility to these conditions.

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSLP
Applications Functional Assay, Neutralization
Classification Monoclonal
Clone eBio28F12 (28F12)
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene TSLP
Gene Accession No. Q9JIE6
Gene Alias Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP
Gene Symbols TSLP
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53603
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.